Learn More
Thermo Scientific™ Medifuge™ Small Benchtop Centrifuge

Perform both routine clinical and life science separations with this compact centrifuge and the unique DualSpin rotor featuring a 2-in-1 hybrid design.
$4,901.17 - $5,016.56
Specifications
| Capacity | 8 x 15 mL (Fixed Angle), 8 x 10 mL (Swinging Bucket) |
|---|---|
| Max. Speed | 4,900 rpm |
| Max. RCF | 3,114 x g (Fixed Angle); 3,490 x g (Swinging Bucket) |
| Refrigerated | No |
| Display | Large, Bright LED |
| Catalog Number | Mfr. No. | Electrical Requirements | Description | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Electrical Requirements | Description | Price | Quantity & Availability | ||||
13101501
![]() |
Thermo Scientific™
75008801 |
120 V 60 Hz | Medifuge Centrifuge |
Each for $4,901.17
|
|
||||
13101500
![]() |
Thermo Scientific™
75008800 |
220/230 V 50/60 Hz | Medifuge Centrifuge |
Each for $5,016.56
|
|
||||
Description
Perform both routine clinical and life science separations with the Thermo Scientific™ Medifuge™ Clinical Centrifuge. It fits in with its compact design and stands out with the unique Thermo Scientific™ DualSpin™ rotor featuring a 2-in-1 hybrid design. Easily change between fixed angle and swinging buckets using the same rotor. Four customizable programs allow you to quickly run routine protocols. Three selectable deceleration profiles, including brake-off option, are designed to optimize separation.
Package content
- Each Medifuge centrifuge comes as a package, including:
- Medifuge small benchtop centrifuge
- DualSpin rotor body
- Fixed angle buckets (set of 8)
- Swinging buckets (set of 8)
- Green/short spacers (set of 8)
- Yellow/long spacers (set of 8)
Applications
- Platelet-poor plasma (PPP) separation
- Urinalysis
- Clinical chemistry analysis
- Hematology analysis
- Serum preparation using gel tube
- Cell culture pelleting
Features
- Unique DualSpin rotor with interchangeable fixed angle and swinging buckets accommodates 1.4 mL to 15 mL tubes with two spacer options
- The lightweight composite 8-place rotor is easy to handle and saves energy during acceleration/deceleration
- Large, brightly-lit display is easy to read and fast one-click lid closure simplifies operation
- Four customizable programs to quickly run routine protocols
- Three selectable deceleration profiles including brake-off option are designed to optimize separation
- Quiet operation at noise level less than 56 dBA
- Emergency lid lock release available in case of power failure
Certifications
UL, CE, RoHS, CSA, IVD, FDA class 1
Standards
IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, EN ISO 13485
Previous models
- Labofuge centrifuge
- Medilite centrifuge
Specifications
| 8 x 15 mL (Fixed Angle), 8 x 10 mL (Swinging Bucket) | |
| 3,114 x g (Fixed Angle); 3,490 x g (Swinging Bucket) | |
| Large, Bright LED | |
| 450 mm | |
| Lid lock, steel armored ring, view port for speed calibration, lid emergency release | |
| 130 w | |
| DualSpin rotor body, fixed angle buckets (set of 8), swinging buckets (set of 8), green/short spacers (set of 8), yellow/long spacers (set of 8) | |
| Microprocessor | |
| Induction drive | |
| Yes, 4 program memory | |
| 2°C to 40°C | |
| 1 accel (standard)/3 decel (standard, soft, brake-off) |
| 4,900 rpm | |
| No | |
| 240 mm | |
| <56 dB(A) (swinging bucket) | |
| 325 mm | |
| UL, CE, RoHS, CSA, IVD, FDA class 1 | |
| Medifuge Centrifuge | |
| 9.5 x 12.8 x 17.7 in. (24 x 32.5 x 45 cm) | |
| 34.1 lb. (15.5 kg) | |
| 1-99 minutes, in increments of 1 minute; HOLD | |
| Medifuge | |
| 510 mm |