missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Rubber Cap/Liner

Catalog No. 06414
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Quantity:
24 EA
48 EA
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
06414 125 mL 48 EA
06415 250 mL 48 EA
22263008 500 mL 24 EA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06414 Supplier DWK Life Sciences Supplier No. 219575

Clear Type I borosilicate glass bottle without graduations.

  • Manufactured from borosilicate glass which provides the best chemical resistance
  • Clear borosilicate glass bottle
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black Phenolic Rubber Lined Cap
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse

Specifications

Capacity (Metric) 125 mL
Color Clear
Material Borosilicate Glass (Type I)
Quantity 48 EA
Closure Color Black
Product Type Media Bottle
Closure Liner Rubber
Closure Material Phenolic