missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, PE Cap/Liner
Clear Type I borosilicate glass bottle without graduations.
$770.91 - $1,161.38
Specifications
| Color | Clear |
|---|---|
| Material | Borosilicate Glass (Type I) |
| Closure Color | Black |
| Product Type | Media Bottle |
| Closure Liner | Polyethylene |
| Catalog Number | Mfr. No. | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
|
06411
|
DWK Life Sciences
219495 |
125 mL | 48 EA |
Case of 48 Each for $789.71
|
|
||||
|
06412
|
DWK Life Sciences
219497 |
250 mL | 48 EA |
Case of 48 Each for $1,161.38
|
|
||||
|
06451278
|
DWK Life Sciences
219499 |
500 mL | 24 EA |
Case of 24 Each for $770.91
|
|
||||
Description
- Manufactured from amber borosilicate glass which provides the best chemical resistance
- Clear borosilicate glass bottle
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black Phenolic Polyethylene Lined Cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Specifications
| Clear | |
| Black | |
| Polyethylene |
| Borosilicate Glass (Type I) | |
| Media Bottle | |
| Phenolic |