missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Capless

Catalog No. 06409
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Quantity:
24 EA
48 EA
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
06409 500 mL 24 EA
06407 125 mL 48 EA
06408 250 mL 48 EA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06409 Supplier DWK Life Sciences Supplier No. 219419

Clear Type I borosilicate glass bottle without graduations.

  • Manufactured from borosilicate glass which provides the best chemical resistance
  • Clear borosilicate glass bottle
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black phenolic screw cap can be purchased separately

Specifications

Capacity (Metric) 500 mL
Color Clear
Material Borosilicate Glass (Type I)
Quantity 24 EA
Neck Finish 33-340
Product Type Media Bottle
Closure Size 33-340 mm
Closure Type No Closure