Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Amber
Catalog No. 06451290
Change view
Click to view available options
Capacity (Metric):
125 mL
Catalog No. Capacity (Metric)
06451290 125 mL
1 options
Catalog No. 06451290 Supplier DWK Life Sciences Supplier No. 219885
Only null left
Tinted Type I borosilicate glass bottle without graduations.
  • Manufactured from borosilicate glass which provides the best chemical resistance
  • Amber borosilicate glass bottle provides protection from UV light
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black phenolic screw cap can be purchased separately
Specifications
Capacity (Metric) 125 mL
Color Amber
Material Borosilicate Glass (Type I)
Quantity 48 EA
Product Type Media Bottle
Closure Type Not Included