Learn More
Keystone Cleanroom Products Media Bottle Pipe and Flask Closures
Media Bottle Pipe and Flask Closures are an efficient and consistent means of preparing pharmaceutical production equipment for steam sterilization.
$3,275.27 - $7,096.44
Specifications
Autoclavable Autoclavable
Closure Type Elastic
Color White
Disposable Single-use
Material Tyvek
View More Specs
Products 7
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19999144
View Documents Promotion Details
Keystone Cleanroom Products
10312TW
8 in.
Case of 1000 Each for $7,096.44
Only null left
19999145
View Documents Promotion Details
Keystone Cleanroom Products
10316TW
12 in.
Case of 500 Each for $4,746.12
Only null left
19999146
View Documents Promotion Details
Keystone Cleanroom Products
1033TW*
1 in.
Case of 1000 Each for $3,275.27
Only null left
19999147
View Documents Promotion Details
Keystone Cleanroom Products
1034TW
1.75 in.
Case of 1000 Each for $3,275.27
Only null left
19999148
View Documents Promotion Details
Keystone Cleanroom Products
1035TW*
2.5 in.
Case of 1000 Each for $3,760.50
Only null left
19999149
View Documents Promotion Details
Keystone Cleanroom Products
1037TW*
4 in.
Case of 1000 Each for $4,306.38
Only null left
19999150
View Documents Promotion Details
Keystone Cleanroom Products
1039TW
5.5 in.
Case of 1000 Each for $4,806.76
Only null left
Description
Description
  • Tyvek Media Bottle Pipe and Flask Closures prevent dust and other particulates from entering orifices of any size.
  • Conveniently covers flasks, beakers, containers, media bottles, valves, pipes and hoses
  • Prevents dust and other particulate from entering orifices of any size
  • Autoclavable - can be used during sterilization to reduce time consuming wrapping
  • Breathable, while providing a superior barrier post sterilization
  • Tyvek has the highest microbial barrier properties among all steam sterilization wrapping compatible materials
  • Manufactured and packaged in a Controlled Cleanroom Environment
  • Resists tearing and punctures
  • Eliminates Autoclave Tape Residue
  • Made from 1422A Tyvek Material
  • Latex-free and Talc-Free rubber elastic
  • Hydrophoic
  • Lint-Free
Specifications
Specifications
Autoclavable
White
Tyvek
Yes
Elastic
Single-use
Non-sterile
Cover