missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MED6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564391 Supplier Invitrogen™ Supplier No. PA564391
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LGISWVDSSW IPILNSGSVL DYFSERSNPF YDRTCNNEVV KMQRLTLEHL NQMVGIEYIL LHAQEPILFI IRKQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

MED6 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. And mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.

Specifications

Antigen MED6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Med6
Gene Accession No. O75586
Gene Alias 1500012F11Rik; Activator-recruited cofactor 33 kDa component; ARC33; AU019660; AV213303; CTD-2540L5.5; hMed6; hypothetical protein LOC505293; MED6; Mediator complex subunit 6; mediator of RNA polymerase II transcriptio subunit 6-like protein; mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; NY-REN-28; Renal carcinoma antigen NY-REN-28
Gene Symbols Med6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED6. Recombinant protein control fragment (Product #RP-103671).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less