missing translation for 'onlineSavingsMsg'
Learn More

Mebhydroline 1,5-naphthalenedisulfonate salt, MilliporeSigma™ Supelco™

Catalog No. 111014899
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014899 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014899 Supplier Merck Emd Millipore Supplier No. M52791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 6153-33-9
Physical Form Neat
Linear Formula C19H20N2 · 1/2C10H8O6S2
MDL Number MFCD00083420
Synonym 9-Benzyl-N-methyl-1,2,3,4-tetrahydro-alpha-carboline
Type Analytical Standard
Quantity 1 g
Formula Weight 420.52
Grade Analytical Standard