missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022838
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022838 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022838 Supplier Merck Emd Millipore Supplier No. BP1008
Only null left

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 31431-39-7
Molecular Formula C16H13N3O3
Linear Formula C16H13N3O3
MDL Number MFCD00057872
Synonym 5-Benzoyl-2-benzimidazolecarbamic acid methyl ester; Mebendazol; Methyl N-(5-benzoyl-1 H-benzimidazol-2-yl)carbamate
Type Pharmacopeia and Metrological Institutes Standards
Quantity 100 mg
Formula Weight 295.29
Grade British Pharmacopoeia (BP) Reference Standard