missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MDM2 Monoclonal Antibody (IF2)
GREENER_CHOICE

Catalog No. 182403
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
182403 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 182403 Supplier Invitrogen™ Supplier No. 182403
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This monoclonal antibody reacts specifically with human MDM2 protein. Overexpression of MDM2 protein has been detected in a variety of human tumors, and appears to result from gene amplification, increased transcript level, and enhanced translation. Reactivity with mouse MDM2 protein has not been observed. Expected Staining Pattern: Nuclear.

MDM2 is a ubiquitin ligase for p53 and plays a central role in regulation of the stability of p53. MDM2 is located on chromosome 12 on the q arm. Akt-mediated phosphorylation of MDM2 at Ser166 and Ser186 increases its interaction with p300, allowing MDM2-mediated ubiquitination and degradation of p53. Phosphorylation of MDM2 also blocks its binding to p19ARF, increasing the degradation of p53. MDM2 has also been shown to negatively regulate p53 function. MDM2 binds and inhibits transactivation role played by p53 and overexpression of MDM2 can result in the inactivation of p53 and decrease its tumor suppressor function. Another process by which MDM2 can inactivate p53 is by degrading p53 as the protein also possesses E3 ubiquitin ligase activity. Further, MDM2 plays important roles in apoptosis and cell cycle. MDM2 is over expressed in a wide range of human malignancies including soft tissue carcinomas and breast cancer. In addition to p53, MDM2 is involved in processes of cell cycle, apoptosis, and tumorigenesis through interactions with proteins that include retinoblastoma 1 and ribosomal protein L5. More than 40 different alternatively spliced transcript variants of MDM2 have been isolated from both tumor and normal tissues.

Specifications

Antigen MDM2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IF2
Conjugate Unconjugated
Dilution Concentrate Antibody
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene MDM2
Gene Accession No. Q00987
Gene Alias 1700007J15Rik; AA415488; ACTFS; double minute 2; double minute 2 protein; double minute 2, human homolog of; p53-binding protein; E3 ubiquitin-protein ligase Mdm2; Hdm2; HDMX; HGNC:6973; human homolog of; MDM2; Mdm-2; MDM2 oncogene, E3 ubiquitin protein ligase; Mdm2 p53 binding protein homolog; MDM2 proto-oncogene; MDM2 proto-oncogene, E3 ubiquitin protein ligase; MDM2 variant FB28; Mdm2, p53 E3 ubiquitin protein ligase homolog; Mdm2, transformed 3T3 cell double minute 2, p53 binding protein; MGC5370; MGC71221; oncoprotein Mdm2; OTTHUMP00000183490; OTTHUMP00000183492; OTTHUMP00000183494; OTTHUMP00000183495; OTTHUMP00000183496; OTTHUMP00000183497; p53 E3 ubiquitin protein ligase; p53-binding protein Mdm2; RING-type E3 ubiquitin transferase Mdm2; transformed mouse 3T3 cell double minute 2
Gene Symbols MDM2
Host Species Mouse
Immunogen Synthetic peptide derived from the n-terminal region of human MDM2.
Purification Method Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Clinical
Primary or Secondary Primary
Gene ID (Entrez) 4193
Packaging 1 Each
Test Specificity MDM2
Includes 0.1% Sodium azide
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Host abrv Ms
Reactivity Human
Shipping Condition Wet Ice
Reactivity abrv Hu
Target Gene MDM2
Validated Application abrv IHC (FFPE)
Show More Show Less