missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MDA5 Recombinant Rabbit Monoclonal Antibody (23GB6270)
GREENER_CHOICE

Catalog No. MA553010
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553010 100 μL
MA553011 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553010 Supplier Invitrogen™ Supplier No. MA553010
Only null left

Rabbit Recombinant Monoclonal Antibody

MDA5 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.

Specifications

Antigen MDA5
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6270
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFIH1
Gene Accession No. Q8R5F7, Q9BYX4
Gene Alias 9130009C22Rik; AGS7; CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; helicard; Helicase with 2 CARD domains; Hlcd; IDDM19; IFIH1; interferon induced with helicase C domain 1; interferon induced with helicase C domain protein 1; interferon induced, with helicase C domain 1; interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA5; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RGD1560496; RH116; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116; SGMRT1
Gene Symbols IFIH1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MDA5 (950-1025AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499801, 64135, 71586
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less