missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCP-1 ProQuantum Immunoassay Kit
GREENER_CHOICE

Catalog No. A35598
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35598 96 Tests
A355985 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A35598 Supplier Invitrogen™ Supplier No. A35598

ELISA

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.

Specifications

Accession Number P13500
Assay Range 0.064 to 5000
Assay Sensitivity <0.05 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene ID (Entrez) 6347
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Storage Requirements Frozen at -20°C
Target CCL2 (MCP-1)
Test Time 30 mins
Total Assay Time 2 hrs
Show More Show Less