missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCM7 Recombinant Rabbit Monoclonal Antibody (23GB930)
GREENER_CHOICE

Catalog No. MA554062
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554062 100 μL
MA554063 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554062 Supplier Invitrogen™ Supplier No. MA554062

Rabbit Recombinant Monoclonal Antibody

MCM7 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

MCM7 encodes a protein that is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme.The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumorsuppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen MCM7
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB930
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mcm7
Gene Accession No. P33993
Gene Alias AI747533; CDC47; CDC47 homolog; CDC47 homolog A; CDC47p; DNA replication licensing factor MCM7; DNA replication licensing factor mcm7-A; DNA replication licensing factor MCM7-like protein; homolog of S. cerevisiae Cdc47; mCDC47; MCM2; MCM7; MCM7 minichromosome maintenance deficient 7; mcm7.S; mcm7-a; mcm7-b; Mcmd7; mini chromosome maintenance deficient 7; minichromosome maintenance complex component 7; minichromosome maintenance complex component 7 S homeolog; minichromosome maintenance deficient 7; minichromosome maintenance deficient 7 (S. cerevisiae); minichromosome maintenance protein 7; minichromosome maintenance protein 7-A; P1 P85MCM; P1.1-MCM3; P1CDC47; P85MCM; p90; PNAS146; PPP1R104; protein phosphatase 1, regulatory subunit 104; Unknown (protein for MGC:128845); XELAEV_18000877mg; XMCM7; xMCM7-A
Gene Symbols Mcm7
Host Species Rabbit
Immunogen A synthetic peptide of human MCM7 (1-50AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288532, 4176
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less