missing translation for 'onlineSavingsMsg'
Learn More

McIlvaine's Buffer, Certified, LabChem™

Catalog No. p-5388056
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC163004 4 L
LC163006 10 L
LC163005 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC163004 Supplier LabChem Supplier No. LC163004
Only null left

Specifications

Chemical Name or Material McIlvaine's Buffer
Color Colorless
CAS Max % 97.74, 1.55, 0.7, 0.01
CAS 77-92-9, 7558-79-4, 89-83-8
Decomposition Information Phosphorus oxides; Carbon dioxide; Carbon monoxide
Density 1g/mL
pH 3.20 ±0.05
Physical Form Liquid
Quantity 4 L
Identification Passes Test
Linear Formula Na2HPO4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 141.96
Grade Certified
Show More Show Less