missing translation for 'onlineSavingsMsg'
Learn More
Learn More
McIlvaine's Buffer, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | McIlvaine's Buffer |
| Color | Colorless |
| CAS Max % | 97.74, 1.55, 0.7, 0.01 |
| CAS | 77-92-9, 7558-79-4, 89-83-8 |
| Decomposition Information | Phosphorus oxides; Carbon dioxide; Carbon monoxide |
| Density | 1g/mL |
| pH | 3.20 ±0.05 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Identification | Passes Test |
| Show More |