missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MC2R Polyclonal Antibody
MC2R Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA525454
Description
MC2R Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| MC2R | |
| Polyclonal | |
| Unconjugated | |
| MC2R | |
| ACTH receptor; ACTHR; ACTH-R; Adrenocorticotropic hormone receptor; adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2R; MC2-R; melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); melanocortin receptor 2; MGC125798 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 4158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q01718 | |
| MC2R | |
| KLH conjugated synthetic peptide between 261-288 amino acids of human MC2R. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |