missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MBP Bovine Protein, Natural

Catalog No. 13228010
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13228010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13228010 Supplier Gibco™ Supplier No. 13228010

missing translation for 'validMainframeMsg'

MBP Bovine Protein, Natural

Myelin Basic Protein (MBP) was purified from bovine brain by a modification of the method of Deibler et al. (1). The protein is supplied lyophilized.

Specifications

Quantity 10 mg
Species Bovine
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Approved for shipment on Wet or Dry Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.