| Accession Number |
P11226 |
| Assay Range |
0.034-25 ng/mL |
| Assay Sensitivity |
0.03 ng/mL |
| Calibration Range |
0.03 ng/mL to 25 ng/mL |
| Content And Storage |
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Assay |
Solid-phase sandwich ELISA |
| Conjugate |
HRP |
| Immunoassay Kit Format |
Sandwich ELISA Kit |
| Product Type |
ELISA |
| Protein Family |
Other Proteins |
| Sample Type |
Plasma, Serum, Supernatant |
| Specificity |
This ELISA antibody pair detects human MBL. Other species not determined yet. |
| Sufficient For |
96 assays |
| Target Species |
Human |
| For Use With (Application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| For Use With (Equipment) |
Absorbance Microplate Reader |
| Gene Alias |
27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin; COLEC1; HSMBPC; MBLD; MBP; MBP-C; MBP1; MBPD; MBL2 |
| Gene ID (Entrez) |
4153 |
| Gene Symbol |
MBL2 |
| Interassay CV |
<12% |
| Intraassay CV |
<10% |
| Kit Contents |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Label or Dye |
Biotin |
| Protein Aliases |
MBP-C, Collectin-1, MBP1, Mannan-binding protein, Mannose-binding lectin |
| Quantity |
96 Tests |
| Regulatory Status |
RUO |
| Research Discipline |
Microbiology |
| Sample Volume |
Plasma 1 μL, Serum 1 μL, Supernatant 100 μL |
| Storage Requirements |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonym |
RP11-94H3.1, COLEC1, HSMBPC, MBLD, MBP, MBP-C, MBP1, MBPD, MBL2 |
| Target |
MBL2 |
| Test Time |
1 hr 20 min |
| Total Assay Time |
4 hr 45 min |
|
Show More
Show Less
|
|