missing translation for 'onlineSavingsMsg'
Learn More

Ohaus MB32 Moisture Analyzer

Catalog No. 01670761
Change view
Click to view available options
Readability:
1 mg/0.01%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Readability
01670761 1 mg/0.01%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01670761 Supplier Ohaus Supplier No. OHS30971928

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Ohaus MB32 moisture analyzer features innovative carbon fiber heating technology and an upgraded load cell for enhanced durability and performance during routine testing.

  • An LCD with second line display, navigation knob, and intuitive operation system makes for convenient routine moisture testing.
  • The new carbon fiber heating technology and upgraded loadcell shortens analysis time, saves power and keeps results accurate and consistent.
  • Robust, slim design with a strong housing ensures MB32's durability even in harsh environments.

Specifications

Readability 1 mg/0.01%
Display Type LCD
Capacity 90 g
Sample Size 3.5 in. (90 mm)
Type Moisture Analyzer
Voltage 120 V