missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Maximum Durability Absorbent Mat Roll

Avoid slips, trips, and falls with this great addition to your absorbent program. The Brady™ Adhesive Mat Roll adheres to any surface and features an easily removed absorbent that won't leave residue on your floor. For use in walkways, areas of heavy traffic and in place of traditional rugs.

$549.50

Specifications

Absorbancy 8 gl/gs
Color Black
Description Needle-punched polypropylene with adhesive backing
Material Polypropylene
For Use With (Application) Walkways, areas of heavy traffic and in place of traditional rugs
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (English) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Dimensions (L x W) Price Quantity & Availability  
19103109
View Documents Promotion Details
Brady™
AD30100
21 gal. 100 ft. x 30 in.
Each for $549.50
Only null left
 
Description

Description

  • Great addition to your absorbent program
  • Tacky underside adheres to any surface, keeping the absorbent in place to greatly reduce the likelihood of slips, trips, and falls
  • When discarding, the absorbent is easily removed without leaving any residue on your floor
  • Mat is barrier-backed and fire resistant
  • For use in walkways, aisles, near equipment and as a replacement for traditional rugs
Specifications

Specifications

8 gl/gs
Needle-punched polypropylene with adhesive backing
Walkways, areas of heavy traffic and in place of traditional rugs
100 ft.
Black
Polypropylene
1 Roll
Videos
SDS
Documents

Documents