missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima Reverse Transcriptase (200 U/μL)

Catalog No. FEREP0741
Change view
Click to view available options
Quantity:
10,000 Units
2,000 Units
4 x 10,000 Units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0741 2,000 Units
FEREP0742 10,000 Units
FEREP0743 4 x 10,000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FEREP0741 Supplier Thermo Scientific™ Supplier No. EP0741

Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity as well as RNase H activity.

Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity as well as RNase H activity. The engineered enzyme features dramatically improved thermostability and robustness, and increased synthesis rates compared to wild type M-MuLV RT.

Maxima Reverse Transcriptase is capable of reproducible cDNA synthesis from a wide range of input total RNA amounts (from 1 pg to 5 μg) at elevated temperatures (50–65°C), which makes this enzyme an ideal tool for two step RT-qPCR (see supporting data).

Due to its high thermostability, Maxima enzyme maintains full activity during the entire reverse transcription reaction, generates the highest yields of cDNA, and is able to synthesize even very long RNA transcripts up to 20 kb. The reaction temperature can be increased up to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. Due to its increased synthesis rate, the reverse transcriptase reaction can be completed in 15 to 30 minutes.

Features of Maxima Reverse Transcriptase include:

  • High yields of full-length cDNA up to 20 kb
  • Active up to 65°C
  • Thermostable—90% active after incubation at 50°C for 60 minutes
  • High sensitivity allows reproducible cDNA synthesis from a wide range of total RNA quantities (1 pg to 5 μg)
  • Efficient formula for complete cDNA synthesis in 15 to 30 minutes
  • Incorporates modified nucleotides

Applications

  • Full-length cDNA synthesis for long transcripts and ORFs
  • Reverse transcription at elevated temperatures to reduce effects of secondary structure
  • cDNA cloning and construct generation
  • RNA-seq & single cell RNA-seq library prep
  • Isoform and transcript characterization
  • Template-switching-based full-length RNA-seq

Also available

  • Maxima Reverse Transcriptase (Cat. No. EP0741)
  • Maxima First Strand cDNA Synthesis Kit for RT-qPCR (Cat. No. K1641)
  • Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase (Cat. No. K1671)

Specifications

Concentration 200 U/μL
Content And Storage

• Maxima Reverse Transcriptase, 1 x 2,000 units (200 U/μL)
• 5X RT Buffer

Store at –20°C.

Format Stand-alone Enzyme
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C to 55°C
Quantity 2,000 Units
Reverse Transcriptase Maxima
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
Final Product Type First-Strand cDNA
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Show More Show Less
When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

Is it preferable to use elevated temperature in the RT reaction when using Maxima reverse transcriptases?

cDNA synthesis at higher temperatures ensures successful transcription of RNA with high levels of secondary structure, reducing issues of primer access to template. Therefore, we do recommend to use RT enzymes with high thermostability, e.g. Maxima and Maxima H Minus Reverse Transcriptases, which provide higher yields of full-length cDNA, better sensitivity, and successful transcription of GC-rich templates.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.


For Research Use Only. Not for use in diagnostic procedures.