Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
GREENER_CHOICE
Catalog No. 14387065
Change view
Click to view available options
Capacity (Metric):
25 mL
75 mL
100 mL
Material:
Polypropylene
Polystyrene
Type:
Reagent Reservoir
Reagent Reservoir with Divider
Reservoir Lid
Packaging:
10 Bags of 10
10 pcs /bag
50 Individually wrapped
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Type Packaging
14387065 75 mL Polypropylene Reagent Reservoir -
50824108 75 mL Polypropylene Reservoir Lid 10 pcs /bag
14387068 100 mL Polystyrene Reagent Reservoir 50 Individually wrapped
14387069 25 mL Polystyrene Reagent Reservoir 10 Bags of 10
14387070 25 mL Polystyrene Reagent Reservoir 50 Individually wrapped
14387071 25 mL Polystyrene Reagent Reservoir with Divider 10 Bags of 10
14387072 25 mL Polystyrene Reagent Reservoir with Divider 50 Individually wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14387065 Supplier Thermo Scientific™ Supplier No. 8075
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Specifications
Color White
Capacity (Metric) 75 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 10
Autoclavable Autoclavable