missing translation for 'onlineSavingsMsg'
Learn More

Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals

Catalog No. p-7052193
Change view
Click to view available options
Quantity:
1 gal.
3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44012EQC 3.8 L
AA44012LV 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44012EQC Supplier Thermo Scientific Chemicals Supplier No. 044012EQC
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64742-65-0
MDL Number MFCD00147539

Specifications

Odor Petroleum-like
Physical Form Liquid
Quantity 3.8 L
Chemical Name or Material Matrix oil, 20 centistoke hydrocarbon oil
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 265-169-7
RTECSNumber PY8038500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only