missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 2.0% Mg (20,000 ppm Mg), Ricca Chemical

Supplier:  Ricca Chemical Company MMMG20KN100

 View more versions of this product

Catalog No. MMMG20KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

76.0, 16.09, 5.72
79.0, 16.74, 5.96
13446-18-9, 7697-37-2
Matrix Modifier

Specifications

Matrix Modifier
2.0% Pd,20,000ppm Pd
79.0, 16.74, 5.96
Natural LDPE Bottle
76.0, 16.09, 5.72
13446-18-9 , 7697-37-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions