missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 1.0% Pd (10,000 ppm Pd), Ricca Chemical

Catalog No. MMPD10KN50
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMPD10KN50 50 mL Natural Poly Bottle
MMPD10KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMPD10KN50 Supplier Ricca Chemical Company Supplier No. MMPD10KN50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
Concentration or Composition Notes 1.0% Pd,10,000ppm Pd
CAS Min % 88.0, 9.23, 0.88, 0.16
CAS Max % 91.0, 9.61, 0.91, 0.16
CAS 7697-37-2, 7440-05-3, 7647-01-0
Physical Form Liquid
Color Brown
Quantity 50 mL