missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 0.2% Pd (2000 ppm Pd), Ricca Chemical

Catalog No. MMPD2KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMPD2KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMPD2KN100 Supplier Ricca Chemical Company Supplier No. MMPD2KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural LDPE Bottle
Concentration or Composition Notes 0.2% Pd,2000ppm Pd
CAS Min % 92.0, 6.16, 0.15, 0.03
CAS Max % 95.0, 6.42, 0.15, 0.03
CAS 7697-37-2, 7440-05-3, 7647-01-0
Physical Form Liquid
Color Brown
Melting Point Approximately 0°C
Quantity 100 mL