missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MATK (Hyl), His Tag Recombinant Protein
Recombinant Protein
$13030.00 - $13030.00
Specifications
| Accession Number | P42679 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 20mM HEPES with 0.5mM EDTA, 5mM DTT, 150mM NaCl, 0.05% CHAPS, 0.5mM EGTA, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 4145 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV4084
|
Invitrogen™
PV4084 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.Specifications
| P42679 | |
| Kinase Assay | |
| 4145 | |
| Human MATK (Hyl), His Tag | |
| Purified | |
| RUO | |
| MATK | |
| CSK Kinase Family | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Tyrosine Kinases | |
| E. coli |
| See Label | |
| 20mM HEPES with 0.5mM EDTA, 5mM DTT, 150mM NaCl, 0.05% CHAPS, 0.5mM EGTA, 50% glycerol and no preservative; pH 7.5 | |
| 53 kDa | |
| 7.5 | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase | |
| MATK | |
| Protein | |
| Human | |
| His-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Dry Ice | |
| Liquid |