missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MATK (Hyl), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
Specifications
Specifications
| Accession Number | P42679 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 20mM HEPES with 0.5mM EDTA, 5mM DTT, 150mM NaCl, 0.05% CHAPS, 0.5mM EGTA, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 4145 |
| Molecular Weight (g/mol) | 53 kDa |
| Name | Human MATK (Hyl), His Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |