Learn More
Brady™ Material Handling, Shipping & Receiving Labels - Self-Sticking Polyester
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
Supplier:  Brady™ 60257
 View more versions of this product
Catalog No. 19-110-988
Only null left
Explore available promotions
Description
Description
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Specifications
Not Autoclavable
Label
Red on Yellow
Label
Single-side Printable
USED OIL
25/Pk.
7 x 3 in. (W x H)
7 in.
3 in.
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Permanent Acrylic, Self-adhesive
Gloss
Polyester
Label
English
Rectangle
3 x 7 in.
85.09 mm
Product Suggestions
Product Suggestions