missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Mastoparan, Calbiochem™,

Catalog No. 4448981MG
Change view
Click to view available options
Quantity:
1 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4448981MG 1 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4448981MG Supplier MilliporeSigma™ Supplier No. 4448981MG

Chemical Identifiers

CAS 72093-21-1
Molecular Formula C70H131N19O15
Molecular Weight (g/mol) 1478.94
MDL Number MFCD00076865
InChI Key MASXKPLGZRMBJF-HDSAQESISA-N
Synonym mastoparan, mastoparan, vespula lewisii, inlkalaalakkil-nh2, mastoparan, vespula lewisii hplc , powder, ile-asn-leu-lys-ala-leu-ala-ala-leu-ala-lys-lys-ile-leu-nh2, l-isoleucyl-l-asparaginyl-l-leucyl-l-lysyl-l-alanyl-l-leucyl-l-alanyl-l-alanyl-l-leucyl-l-alanyl-l-lysyl-l-lysyl-l-isoleucyl-l-leucinamide
PubChem CID 6324633
ChEBI CHEBI:78496
IUPAC Name (2S)-N-[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-carbamoyl-3-methylbutyl]carbamoyl}-2-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}pentyl]carbamoyl}-3-methylbutyl]-2-[(2S,3S)-2-amino-3-methylpentanamido]butanediamide
SMILES CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)CC)C(=O)N[C@@H](CC(C)C)C(N)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 1 mg
Physical Form Solid