accessibility menu, dialog, popup

UserName

H3C(S)CH3(S)NH2OHN(S)NH2OONH(S)CH3CH3OHN(S)NH2ONH(S)CH3OHN(S)CH3CH3ONH(S)CH3OHN(S)CH3ONH(S)CH3CH3OHN(S)CH3ONH(S)NH2OHN(S)NH2ONH(S)H3CCH3OHN(S)CH3CH3NH2O

CAS RN 72093-21-1

IUPAC Name: (2S)-N-[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-carbamoyl-3-methylbutyl]carbamoyl}-2-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}pentyl]carbamoyl}-3-methylbutyl]-2-[(2S,3S)-2-amino-3-methylpentanamido]butanediamide
Synonyms: (2S)-N-[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-carbamoyl-3-methylbutyl]carbamoyl}-2-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}pentyl]carbamoyl}-3-methylbutyl]-2-[(2S,3S)-2-amino-3-methylpentanamido]succinamide
Molecular Weight (g/mol): 1478.94
Molecular Formula: C70H131N19O15
InChi Key: MASXKPLGZRMBJF-HDSAQESISA-N
SMILES: CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)CC)C(=O)N[C@@H](CC(C)C)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results