Learn More
StatLab MasterTech Myofibrillar ATPase Stain Kit
Description
- StatLab Myofibrillar ATPase Stain Kit contains no barbiturates - In the Myofibrillar ATPase Stain Kit a predominance of Type 1 fiber types typically indicates a myopathic condition, such as dystrophy, while a predominance of Type 2 fibers typically indicates a neuropathic condition
- Type 1 Buffer stains small type 1 fibers black and type 2C fibers gray
- Type 1 and 2BC Buffer stains small type 1 fibers black, 2B and 2C fibers gray to black
- Type 2AB Buffer stains type 2A and 2B fibers black
- 100 mL Stain Kit yields between 36-45 slides
- 100 mL Kit Consists of: Type 1 Buffer 100 mL, Type 1 and 2BC Buffer 100 mL, Alkaline Buffer Stock 100 mL x 2, Calcium Chloride Solution 100 mL, Cobalt Chloride Solution 100 mL, 1N Sodium Hydroxide 15 mL, 1N Hydrochloric Acid 15 mL, ATP Powder 200 mg
- A MasterTech Special Stain Kit
Spécifications
Spécifications
| Méthode de détection | Chromogenic |
| Format de la trousse d’immunodosage | Other Format |
| Type de produit | Special Stain Kit |