Learn More
Greiner Bio-One MASTERBLOCK™ 384 Deep Well Polypropylene Conical Bottom Microplates
Free of detectable DNase, RNase, human DNA.
$1,372.75 - $1,389.29
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Non Binding
Color Clear
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
07000048
View Documents Promotion Details
Greiner Bio-One
781270
Non-sterile
Case of 60 Each for $1,372.75
Only null left
07000885
View Documents Promotion Details
Greiner Bio-One
781271
Sterile
Case of 60 Each for $1,389.29
Only null left
Description
Description
  • Alphanumeric well coding
  • High chemical resistanceHigh temperature resistanceSealable with adhesive films and heat sealer
Specifications
Specifications
Standard
16 x 24
Clear
384
240 μL
6
Without Lid
Non Binding
Polypropylene
20 to 225 μL
Rounded Square, Conical Bottom (V-well)
Clear