Learn More
Master Lock™ Safety Lockout Hasp DFS Item
Allows up to six workers to lock out a single energy source
Supplier:  Master Lock™ 421
 View more versions of this product
Catalog No. 29-635-9300
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Heavy gauge steel jaw with red vinyl coated handle
  • Accepts padlocks up to .28 in shackle diameter
  • Control cannot be turned on until last workers padlock is removed from hasp
Specifications
Specifications
Red
0.37 in.
0.93 cm
To lock out a single energy source
13 cm
6 cm
1.5 in.
4 cm
5 x 2.37 in. (13 x 6 cm)
5 in.
2.37 in.
Product Suggestions
Product Suggestions