Learn More
Master Lock™ Safety Lockout Hasp
Allows up to six workers to lock out a single energy source
Supplier: Master Lock™ 420
Description
- Heavy gauge steel jaw with red vinyl coated handle
- Accepts padlocks up to .28 in shackle diameter
- Control cannot be turned on until last workers padlock is removed from hasp
Specifications
| Red | |
| 0.37 in. | |
| 0.93 cm | |
| To lock out a single energy source | |
| 11 cm | |
| 4 cm |
| 1 in. | |
| 3 cm | |
| 4.5 x 1.75 in. (11 x 4 cm) | |
| 4.5 in. | |
| 1.75 in. |