missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
GREENER_CHOICE

Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.

$218.12 - $257.12

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Bromophenol Blue
Ready to Load Yes
Gel Compatibility Agarose Gels
View More Specs

Products 5
Catalog Number Mfr. No. Size Range Price Quantity & Availability  
Catalog Number Mfr. No. Size Range Price Quantity & Availability  
FERSM0383
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM0383
80 to 1031 bp
Each for $230.06
Only null left
 
FERSM0403
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM0403
80 to 10000 bp
Each for $218.12
Only null left
 
FERSM1263
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM1263
100 to 1000 bp
Each for $220.83
Only null left
 
FERSM1283
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM1283
100 to 1000 bp
Each for $257.12
Only null left
 
FERSM0393
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM0393
1500 to 10000 bp
Each for $239.48
Only null left
 
Description

Description

Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.

Highlights

  • Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

Specifications

Specifications

6X
Bromophenol Blue
Agarose Gels
MassRuler
Agarose
Yes
2 x 500 μL
Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 Months when stored at room temperature.

SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.