Learn More
Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
GREENER_CHOICE
Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.
Supplier:  Thermo Scientific™ SM1263
 View more versions of this product
Catalog No. FERSM1263
Only null left
Description
Description
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
  • Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
Chemical Identifiers
6X
Bromophenol Blue
Agarose Gels
MassRuler
Agarose
Yes
2 x 500 μL
Specifications
6X
Bromophenol Blue
100 to 1000 bp
2 x 500 μL
DNA Ladder
Agarose
Yes
Agarose Gels
MassRuler
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 24 months when stored at room temperature.
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.