Learn More
Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.
Supplier: Thermo Scientific™ SM1283
Description
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
- Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
| 6X | |
| Bromophenol Blue | |
| 100 to 1000 bp | |
| 2 x 500 μL | |
| DNA Ladder Mix |
| Agarose | |
| Yes | |
| Agarose Gels | |
| MassRuler |
For Research Use Only. Not for use in diagnostic procedures.