missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ MarvelX™ UHPLC Connection Systems PEEK Lined Stainless Steel Tubing
Expertly designed for easier routing throughout instruments, with consistent performance and superior reusability
Supplier: MilliporeSigma™ Supelco™ 51875U
Description
- Built with convenient, removable stainless steel fitting and changeable precision-cut flexible tubing
- Enhanced proprietary tip design ensures zero dead volume (ZDV)
- Compatible with 10-32 coned receiving ports
- Seals up to 19,000 psi (1,310 bar)
- Requires only 2 in-lbs of installation torque
- Superior re-usability, can be used up to 200 times
- Consistent performance
- Truly finger-tight - no tool required
Chemical Identifiers
| 10-32 Coned Receiving Ports | |
| Tubing, Stainless Steel Finger Tight Fittings (2) | |
| Stainless Steel |
| Tubing | |
| PEEK-lined | |
| 19,000 psi (1,310 bar) |
Specifications
| 10-32 Coned Receiving Ports | |
| Tubing, Stainless Steel Finger Tight Fittings (2) | |
| 100 μm | |
| Stainless Steel |
| Tubing | |
| PEEK-lined | |
| 150 mm | |
| 19,000 psi (1,310 bar) |
MarvelX is a trademark of IDEX Health & Science, LLC