missing translation for 'onlineSavingsMsg'
Learn More

MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 Mouse Monoclonal Antibody, Epredia™

Epredia™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Thermo Fisher Scientific MS716P1ABX

 View more versions of this product

Catalog No. MS716P1ABX


Only null left

Description

Description

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10 + M2-9E3

Isotype: IgG2b/κ + IgG2b/κ

Species Reactivity: Human and Horse. Others not known.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Chemical Identifiers

MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Recombinant hMART-1 protein
M2-7C10 + M2-9E3
Monoclonal
Mouse
Cancer and Tumor Biology
Primary

Specifications

MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3
Monoclonal
1mg/mL
Mouse
100 μL
Cancer and Tumor Biology
Equine, Human
IgG2b κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
M2-7C10 + M2-9E3
Unconjugated
Recombinant hMART-1 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions