Learn More
Invitrogen™ MARK4 Monoclonal Antibody (OTI9B7)
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA527002
Catalog No. PIMA527002
Only null left
Description
Description
MARK4 Monoclonal Antibody for Western Blot, IHC (P)
MARK4 is a serine/threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells. The encoded protein is associated with the centrosome throughout mitosis and may be involved in cell cycle control. Expression of MARK4 is a potential marker for cancer, and the encoded protein may also play a role in Alzheimer's disease. Pseudogenes of MARK4 are located on both the short and long arm of chromosome 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for MARK4.
Specifications
Chemical Identifiers
MARK4
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96L34
57787
Human recombinant protein fragment corresponding to amino acids 390-467 of MARK4 produced in E.coli.
Antibody
OTI9B7
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG1
Human
Mark4
2410090P21Rik; C79806; FLJ90097; KIAA1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D
Mark4
Primary
Affinity Chromatography
Specifications
MARK4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96L34
Mark4
Human recombinant protein fragment corresponding to amino acids 390-467 of MARK4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI9B7
Unconjugated
Mark4
2410090P21Rik; C79806; FLJ90097; KIAA1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D
Mouse
Affinity Chromatography
RUO
57787
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions