missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | MARK3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| MARK3 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| CTAK1, KP78, PAR1A | |
| MARK3 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P27448 | |
| 4140 | |
| Between 525 and 575 | |
| Primary | |
| 4° C | |
| MARK3 |