missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Marimastat, 98%

Supplier:  Thermo Scientific Chemicals J67288MA

 View more versions of this product

Catalog No. AAJ67288MA


Only null left
Explore available promotions

Description

Description

A broad spectrum inhibitor of matrix metalloproteases (MMPs); inhibits MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 values of 3, 5, 6, 9 and 13 nM respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

10 mg
-30°C to -10°C
154039-60-8
331.41g/mol
148°C
White
98%
C15H29N3O5
(2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516
Solid

Chemical Identifiers

154039-60-8
C15H29N3O5
(2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only