missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Marimastat, 98%

Catalog No. p-7057762
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67288MCR 1 mg
AAJ67288MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67288MCR Supplier Thermo Scientific Chemicals Supplier No. J67288MCR

A broad spectrum inhibitor of matrix metalloproteases (MMPs); inhibits MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 values of 3, 5, 6, 9 and 13 nM respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154039-60-8
Molecular Formula C15H29N3O5
Synonym (2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
Formula Weight 331.41g/mol
Melting Point 148°C
Form Solid
Assay 98%

RUO – Research Use Only