missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARCO Recombinant Mouse Monoclonal Antibody (PLK1)

Description
MARCO Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), Flow, IP
Specifications
Specifications
| Antigen | MARCO |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | PLK1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Marco |
| Gene Accession No. | Q9UEW3 |
| Gene Alias | AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6 |
| Show More |