missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Crystal, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18605216
Change view
Click to view available options
Quantity:
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605216 25 kg Fiber Drum
18605214 500 g Amber Glass Bottle
18605215 12 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605216 Supplier Spectrum Chemical Mfg Cor Supplier No. P114525KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1145, 88-89-1, C6H2(NO2)3OH

Spectrum™ Chemical Picric Acid, Crystal, Reagent, ACS , also known as 2,4,6-trinitrophenol or TNP. It is a yellow crystalline solid and one of the most acidic phenols. As a highly nitrated compound, picric acid is highly explosive (like TNT). Apart from it's use as an explosive, it also has some uses in organic chemistry for the preparation of crystalline salts of organic bases. It is also used in clinical chemistry for the Jaffe reaction (a colorimetric method to determine creatinine levels in blood and urine).

Chemical Identifiers

CAS 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) 229.10
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 1
Grade ACS
Quantity 25 kg
Packaging Fiber Drum