missing translation for 'onlineSavingsMsg'
Learn More

Marbofloxacin 98.0+%, TCI America™

Catalog No. M22401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22401G 1 g
M22405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22401G Supplier TCI America Supplier No. M22401G

Chemical Identifiers

CAS 115550-35-1
Molecular Formula C17H19FN4O4
Molecular Weight (g/mol) 362.36
MDL Number MFCD00864820
InChI Key BPFYOAJNDMUVBL-UHFFFAOYSA-N
Synonym marbofloxacin, zeniquin, marbocyl, marbofloxacine, marbofloxacine inn-french, marbofloxacinum inn-latin, marbofloxacino inn-spanish, unii-8x09wu898t, dsstox_cid_26600, dsstox_rid_81755
PubChem CID 60651
IUPAC Name 7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1,2-diazatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
SMILES CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN3N(C)COC1=C23)C(O)=O

Specifications

Melting Point 269°C
Color Yellow
Quantity 1 g
Formula Weight 362.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Marbofloxacin
EINECSNumber (5)-6558
RTECSNumber UU8815140
TSCA No