missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTRAX™ Marble Tuff™ Anti-Fatigue Mat
Marble Tuff™ Anti-Fatigue Floor Mat provides comfort and ergonomic support
Supplier: NOTRAX™ 511S0035BU
Description
- Durable smooth marble-pattern top surface of heavy-duty vinyl
- Easy to sweep clean or damp mop
- Dense closed cell PVC foam base provides added cushioning comfort
- Smooth attractive surface east to sweep clean
- Beveled edges on all four sides
- Thickness: 0.5 in.
Chemical Identifiers
| 1 Ea. | |
| Marble pattern top |
Specifications
| 3 x 5 ft. | |
| 3 ft. | |
| 5 ft. | |
| 1 Ea. |
| 0.5 in. | |
| Blue | |
| Marble pattern top |