missing translation for 'onlineSavingsMsg'
Learn More

NOTRAX™ Marble Tuff™ Anti-Fatigue Mat

Marble Tuff™ Anti-Fatigue Floor Mat provides comfort and ergonomic support

Supplier:  NOTRAX™ 511S0035BL

 View more versions of this product

Catalog No. 19086102


Only null left

Description

Description

  • Durable smooth marble-pattern top surface of heavy-duty vinyl
  • Easy to sweep clean or damp mop
  • Dense closed cell PVC foam base provides added cushioning comfort
  • Smooth attractive surface east to sweep clean
  • Beveled edges on all four sides
  • Thickness: 0.5 in.
Specifications

Chemical Identifiers

1 Ea.
Marble pattern top

Specifications

5 ft.
0.5 in.
3 ft.
1 Ea.
3 x 5 ft.
Marble pattern top
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions