Learn More
NoTrax™ Marble Sof-Tyle™ Anti-Fatigue Mat
Marble Sof-Tyle™ is an anti-fatigue mat with a heavy-duty vinyl stylish smooth marble-pattern design with a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.
$4448.46 - $5931.27
Specifications
Certifications/Compliance Certified by the NFSI (National Floor Safety Institute)
Length (English) 75 ft.
Length (Metric) 22.86 m
Material Nitricell, PVC
Type Anti-fatigue Floor Mat
View More Specs
Products 5
Catalog Number Mfr. No. Dimensions (L x W) Color Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Color Price Quantity  
19-015-808
View Documents
NoTrax™
470R0036BU
75 x 3 ft. Blue
Each for $4,448.46
Only null left
19-015-807
View Documents
NoTrax™
470R0036GY
75 x 3 ft. Smoke Gray
Each for $4,448.46
Only null left
19-015-805
View Documents
NoTrax™
470R0048BU
75 x 4 ft. Blue
Each for $5,931.27
Only null left
19-015-804
View Documents
NoTrax™
470R0048GY
75 x 4 ft. Smoke Gray
Each for $5,931.27
Only null left
19-015-806
View Documents
NoTrax™
470R0048BL
75 x 4 ft. Black Smoke
Each for $5,931.27
Only null left
Description
Description
  • Marble Sof-Tyle™ is an anti-fatigue floor mat provides comfort and ergonomic support, while also complementing any décor with a stylish smooth marble-pattern design
  • The durable vinyl top surface is easy to sweep clean or damp mop and is combined with a dense closed-cell foam base that utilizes NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
  • NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
  • For dry applications such as lab testing area, check-out counters, order fulfillment lines and pharmacies
  • The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat
Specifications
Specifications
Certified by the NFSI (National Floor Safety Institute)
22.86 m
Anti-fatigue Floor Mat
1/2 in.
75 ft.
Nitricell, PVC
Placement in manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual work stations
0.0127 m
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'